Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143197.2 | NP_651454.1 | 1 - 2 | 70 | 72 | |
NM_206576.2 | NP_996299.1 | 1 - 2 | 70 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |