Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143232.3 | NP_651489.2 | 1 | 778 | 67 | |
GenBank mRNA | BT100085.2 | - | 1 | 740 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |