Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276084.1 | NP_001263013.1 | 6 - 7 | 4088 | 79 | |
NM_143314.4 | NP_651571.3 | 6 - 7 | 4088 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |