Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143414.4 | NP_651671.1 | 2 - 3 | 1375 | 83 | |
GenBank mRNA | AY069433.1 | - | 2 - 3 | 1332 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |