Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143452.3 | NP_651709.1 | 3 - 4 | 1576 | 63 | |
GenBank mRNA | AY122183.1 | - | 3 - 4 | 1551 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |