Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143480.4 | NP_651737.1 | 4 - 5 | 1366 | 135 | |
GenBank mRNA | AY061156.1 | - | 4 - 5 | 1346 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |