Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aralar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260439.1 | NP_001247368.1 | 4 - 5 | 631 | 72 | |
NM_001276178.1 | NP_001263107.1 | 4 - 5 | 592 | 72 | ||
NM_143538.3 | NP_651795.2 | 4 - 5 | 592 | 72 | ||
NM_170485.2 | NP_733364.1 | 4 - 5 | 631 | 72 | ||
NM_170486.2 | NP_733365.1 | 4 - 5 | 592 | 72 | ||
NM_170487.3 | NP_733366.2 | 4 - 5 | 767 | 72 | ||
GenBank mRNA | AY058654.1 | - | 4 - 5 | 557 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |