Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aralar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276178.1 | NP_001263107.1 | 6 - 7 | 1006 | 55 | |
NM_143538.3 | NP_651795.2 | 6 - 7 | 1006 | 55 | ||
NM_170485.2 | NP_733364.1 | 6 - 7 | 1045 | 55 | ||
NM_170486.2 | NP_733365.1 | 6 - 7 | 1006 | 55 | ||
NM_170487.3 | NP_733366.2 | 6 - 7 | 1181 | 55 | ||
GenBank mRNA | AY058654.1 | - | 6 - 7 | 971 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |