Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dj-1beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143568.3 | NP_651825.4 | 1 - 2 | 163 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |