Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spn-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276209.1 | NP_001263138.1 | 1 - 2 | 849 | 79 | |
NM_143601.3 | NP_651858.1 | 1 - 2 | 849 | 79 | ||
GenBank mRNA | AY061046.1 | - | 1 - 2 | 818 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |