Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt302K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300352.1 | NP_001287281.1 | 1 - 2 | 295 | 119 | |
NM_144363.2 | NP_652620.1 | 1 - 2 | 295 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |