Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG46339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104469.3 | NP_001097939.1 | 7 - 8 | 2044 | 70 | |
NM_001260394.2 | NP_001247323.1 | 7 - 8 | 1747 | 70 | ||
NM_001276072.1 | NP_001263001.1 | 8 - 9 | 2080 | 70 | ||
NM_001363935.1 | NP_001350863.1 | 5 - 6 | 1615 | 70 | ||
NM_001363936.1 | NP_001350864.1 | 6 - 7 | 1948 | 70 | ||
NM_144487.3 | NP_652744.2 | 6 - 7 | 1711 | 70 | ||
GenBank mRNA | AF480087.1 | - | 6 - 7 | 1688 | 70 | |
AY122083.1 | - | 6 - 7 | 1751 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |