Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4KIIalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382052.1 | NP_001368956.1 | 7 - 8 | 2081 | 93 | |
NM_001382053.1 | NP_001368992.1 | 6 - 7 | 1407 | 93 | ||
NM_169063.4 | NP_730944.2 | 8 - 9 | 2485 | 93 | ||
NM_169064.2 | NP_730945.1 | 6 - 7 | 1928 | 93 | ||
NM_206432.3 | NP_996154.1 | 8 - 9 | 2458 | 93 | ||
GenBank mRNA | AY102687.1 | - | 8 - 9 | 2431 | 93 | |
BT001304.1 | - | 6 - 7 | 1928 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |