Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfzf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014609.3 | NP_001014609.2 | 2 - 3 | 201 | 60 | |
NM_001014610.2 | NP_001014610.1 | 3 - 4 | 2911 | 60 | ||
NM_001275406.1 | NP_001262335.1 | 3 - 4 | 2911 | 60 | ||
GenBank mRNA | AY545463.1 | - | 3 - 4 | 2882 | 60 | |
BT011060.1 | - | 3 - 4 | 2882 | 60 | ||
BT012465.1 | - | 2 - 3 | 198 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |