Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275536.1 | NP_001262465.1 | 5 - 6 | 1274 | 70 | |
NM_169343.4 | NP_731479.3 | 4 - 5 | 1325 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Drosophila melanogaster |