Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169482.1 | NP_731749.3 | 2 - 3 | 482 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |