Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169508.2 | NP_731791.2 | 3 - 4 | 838 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |