Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cv-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104316.2 | NP_001097786.1 | 10 - 11 | 5501 | 63 | |
NM_001104318.2 | NP_001097788.1 | 11 - 12 | 4834 | 63 | ||
NM_169583.3 | NP_731907.1 | 3 - 4 | 681 | 63 | ||
GenBank mRNA | BT044146.1 | - | 3 - 4 | 673 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |