Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cv-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104316.2 | NP_001097786.1 | 13 - 14 | 7908 | 58 | |
NM_001104318.2 | NP_001097788.1 | 14 - 15 | 7241 | 58 | ||
NM_169583.3 | NP_731907.1 | 6 - 7 | 3088 | 58 | ||
GenBank mRNA | BT044146.1 | - | 6 - 7 | 3080 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |