Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169622.3 | NP_731983.1 | 2 - 3 | 418 | 59 | |
GenBank mRNA | BT023312.1 | - | 2 - 3 | 365 | 59 | |
BT029405.1 | - | 2 - 3 | 438 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |