Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104355.2 | NP_001097825.1 | 15 - 16 | 2838 | 56 | |
NM_001104356.2 | NP_001097826.1 | 15 - 16 | 2361 | 56 | ||
NM_169761.2 | NP_732242.2 | 15 - 16 | 2361 | 56 | ||
NM_206504.2 | NP_996226.2 | 15 - 16 | 2361 | 56 | ||
GenBank mRNA | BT014656.1 | - | 15 - 16 | 2309 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |