Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104378.2 | NP_001097848.2 | 11 - 12 | 2297 | 68 | |
NM_001275848.1 | NP_001262777.1 | 11 - 12 | 2070 | 68 | ||
NM_001275849.1 | NP_001262778.1 | 12 - 13 | 2331 | 68 | ||
NM_001275850.1 | NP_001262779.1 | 12 - 13 | 2307 | 68 | ||
NM_001275851.1 | NP_001262780.1 | 11 - 12 | 3632 | 68 | ||
NM_001275853.1 | NP_001262782.1 | 11 - 12 | 2297 | 68 | ||
NM_001275854.1 | NP_001262783.1 | 11 - 12 | 2070 | 68 | ||
NM_001369986.1 | NP_001356920.1 | 12 - 13 | 2307 | 68 | ||
NM_142656.4 | NP_650913.2 | 11 - 12 | 3632 | 68 | ||
NM_169926.2 | NP_732556.1 | 11 - 12 | 2046 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |