Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104392.3 | NP_001097862.1 | 3 - 4 | 1707 | 63 | |
NM_169970.3 | NP_732661.1 | 2 - 3 | 1802 | 63 | ||
NM_169971.3 | NP_732662.2 | 3 - 4 | 1707 | 63 | ||
GenBank mRNA | BT044550.1 | - | 3 - 4 | 1667 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |