Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170032.3 | NP_732775.1 | 2 - 3 | 1176 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |