Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KrT95D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170238.2 | NP_001163709.2 | 6 - 7 | 3176 | 77 | |
NM_079749.6 | NP_524473.2 | 7 - 8 | 3302 | 77 | ||
NM_170108.4 | NP_732933.1 | 8 - 9 | 2622 | 77 | ||
NM_170109.2 | NP_732934.2 | 5 - 6 | 1638 | 77 | ||
NM_170110.4 | NP_732935.2 | 7 - 8 | 2496 | 77 | ||
GenBank mRNA | AY061587.1 | - | 6 - 7 | 1526 | 77 | |
AY119444.1 | - | 7 - 8 | 2479 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |