Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079758.3 | NP_524482.1 | 2 - 3 | 931 | 68 | |
NM_170146.2 | NP_733001.1 | 3 - 4 | 863 | 68 | ||
GenBank mRNA | AY058446.1 | - | 2 - 3 | 917 | 68 | |
X52274.1 | - | 902 | 68 | |||
X53583.1 | - | 753 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |