Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LpR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104456.2 | NP_001097926.1 | 2 - 3 | 795 | 121 | |
NM_001104457.2 | NP_001097927.1 | 2 - 3 | 795 | 121 | ||
NM_001104458.2 | NP_001097928.1 | 2 - 3 | 795 | 121 | ||
NM_001104460.2 | NP_001097930.1 | 2 - 3 | 795 | 121 | ||
NM_170239.4 | NP_733118.3 | 2 - 3 | 795 | 121 | ||
GenBank mRNA | BT010074.1 | - | 2 - 3 | 773 | 121 | |
BT011437.1 | - | 2 - 3 | 796 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |