Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300612.1 | NP_001287541.1 | 3 - 4 | 1275 | 70 | |
NM_170262.2 | NP_733141.1 | 3 - 4 | 1275 | 70 | ||
NM_170263.4 | NP_733142.1 | 3 - 4 | 1053 | 70 | ||
NM_170264.3 | NP_733143.1 | 2 - 3 | 1257 | 70 | ||
NM_170265.4 | NP_733144.3 | 3 - 4 | 1246 | 70 | ||
GenBank mRNA | AY058667.1 | - | 3 - 4 | 1261 | 70 | |
BT099760.1 | - | 3 - 4 | 1048 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |