Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079763.4 | NP_524487.2 | 3 - 4 | 668 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |