Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104472.2 | NP_001097942.1 | 6 - 7 | 1785 | 65 | |
NM_001144644.2 | NP_001138116.1 | 5 - 6 | 1725 | 65 | ||
NM_079802.3 | NP_524526.1 | 5 - 6 | 1893 | 65 | ||
NM_170309.2 | NP_733188.1 | 4 - 5 | 1596 | 65 | ||
NM_170312.2 | NP_733191.1 | 4 - 5 | 1815 | 65 | ||
NM_170313.2 | NP_733192.1 | 5 - 6 | 1647 | 65 | ||
NM_170315.2 | NP_733194.1 | 5 - 6 | 1674 | 65 | ||
NM_170316.2 | NP_733195.1 | 5 - 6 | 1734 | 65 | ||
GenBank mRNA | BT001898.1 | - | 6 - 7 | 1524 | 65 | |
BT044193.2 | - | 4 - 5 | 1797 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |