Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 26 - 27 | 8576 | 90 | |
NM_001276107.1 | NP_001263036.1 | 20 - 21 | 6258 | 90 | ||
NM_079817.3 | NP_524541.3 | 27 - 28 | 8597 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |