Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079862.3 | NP_524601.2 | 2 - 3 | 625 | 84 | |
NM_170534.2 | NP_733413.1 | 2 - 3 | 493 | 84 | ||
GenBank mRNA | BT057987.1 | - | 2 - 3 | 622 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |