Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: disp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079995.3 | NP_524734.2 | 6 - 7 | 3821 | 54 | |
GenBank mRNA | BT050429.1 | - | 6 - 7 | 3821 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |