Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260095.2 | NP_001247024.1 | 2 - 3 | 1509 | 89 | |
NM_001260096.2 | NP_001247025.1 | 3 - 4 | 2093 | 89 | ||
NM_080036.3 | NP_524775.1 | 3 - 4 | 1633 | 89 | ||
GenBank mRNA | AY119588.1 | - | 2 - 3 | 447 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |