Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080175.3 | NP_524914.3 | 1 | 629 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |