Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp1-87B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080198.3 | NP_524937.1 | 1 - 2 | 1312 | 97 | |
GenBank mRNA | AY061063.1 | - | 1 - 2 | 1312 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |