Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: larp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170286.3 | NP_001163757.1 | 3 - 4 | 594 | 72 | |
NM_001260417.2 | NP_001247346.1 | 3 - 4 | 844 | 72 | ||
NM_001260418.2 | NP_001247347.1 | 4 - 5 | 1689 | 72 | ||
NM_001260419.2 | NP_001247348.1 | 4 - 5 | 1693 | 72 | ||
NM_080259.3 | NP_524998.1 | 3 - 4 | 519 | 72 | ||
NM_170365.4 | NP_733244.5 | 4 - 5 | 1838 | 72 | ||
NM_170366.4 | NP_733245.1 | 3 - 4 | 531 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |