Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nonA-l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275717.1 | NP_001262646.1 | 1 | 2232 | 145 | |
NM_080492.3 | NP_536740.1 | 1 | 2232 | 145 | ||
GenBank mRNA | BT150051.1 | - | 1 | 2231 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |