Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276100.1 | NP_001263029.1 | 1 - 2 | 423 | 62 | |
NM_170360.2 | NP_733239.1 | 1 - 2 | 423 | 62 | ||
GenBank mRNA | BT025221.1 | - | 1 - 2 | 419 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |