Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144652.2 | NP_001138124.2 | 3 - 4 | 780 | 89 | |
NM_170434.4 | NP_733313.2 | 3 - 4 | 780 | 89 | ||
GenBank mRNA | BT024267.1 | - | 3 - 4 | 730 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |