Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170455.3 | NP_733334.1 | 5 - 6 | 1140 | 99 | |
GenBank mRNA | AY212802.1 | - | 5 - 6 | 1116 | 99 | |
BT003184.1 | - | 5 - 6 | 1116 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |