Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276169.1 | NP_001263098.1 | 4 - 5 | 1429 | 91 | |
NM_170457.3 | NP_733336.1 | 6 - 7 | 2482 | 91 | ||
NM_170458.2 | NP_733337.1 | 4 - 5 | 1429 | 91 | ||
NM_170459.3 | NP_733338.1 | 6 - 7 | 2482 | 91 | ||
GenBank mRNA | AF091813.1 | - | 6 - 7 | 1752 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |