Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170306.2 | NP_001163777.1 | 2 - 3 | 520 | 63 | |
NM_001202390.1 | NP_001189319.1 | 2 - 3 | 482 | 63 | ||
NM_001260444.2 | NP_001247373.1 | 2 - 3 | 513 | 63 | ||
NM_170506.3 | NP_733385.1 | 2 - 3 | 483 | 63 | ||
GenBank mRNA | BT099728.1 | - | 2 - 3 | 483 | 63 | |
BT100046.1 | - | 2 - 3 | 521 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |