Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144658.2 | NP_001138130.2 | 3 - 4 | 1756 | 91 | |
NM_170531.4 | NP_733410.2 | 3 - 4 | 1756 | 91 | ||
NM_206591.2 | NP_996314.1 | 3 - 4 | 1771 | 91 | ||
GenBank mRNA | BT009990.1 | - | 3 - 4 | 1754 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |