Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170223.1 | NP_001163694.1 | 4 - 5 | 782 | 58 | |
NM_206544.2 | NP_996267.1 | 4 - 5 | 557 | 58 | ||
NM_206545.2 | NP_996268.1 | 3 - 4 | 264 | 58 | ||
GenBank mRNA | AF192555.1 | - | 3 - 4 | 248 | 58 | |
AF224713.1 | - | 3 - 4 | 241 | 58 | ||
AF224714.1 | - | 3 - 4 | 248 | 58 | ||
AF244918.1 | - | 3 - 4 | 248 | 58 | ||
AF250918.1 | - | 3 - 4 | 248 | 58 | ||
AF263722.1 | - | 3 - 4 | 264 | 58 | ||
AY069096.1 | - | 3 - 4 | 248 | 58 | ||
DQ191318.1 | - | 4 - 5 | 497 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |