Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAP-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382065.1 | NP_001368957.1 | 4 - 5 | 1347 | 88 | |
NM_206574.4 | NP_996297.3 | 4 - 5 | 1347 | 88 | ||
GenBank mRNA | BT133202.1 | - | 4 - 5 | 1347 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |