Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170162.2 | NP_001163633.1 | 3 - 4 | 444 | 66 | |
NM_001275725.1 | NP_001262654.1 | 3 - 4 | 765 | 66 | ||
NM_206501.3 | NP_996223.1 | 3 - 4 | 771 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |