Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169757.4 | NP_732230.1 | 1 - 2 | 498 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |