Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258604.2 | NP_001245533.1 | 2 | 1082 | 88 | |
NM_078494.4 | NP_511049.1 | 3 | 1136 | 88 | ||
GenBank mRNA | U25057.1 | - | 3 | 1135 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |