Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: betaTub56D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079071.3 | NP_523795.2 | 2 | 1520 | 143 | |
NM_166356.2 | NP_725896.1 | 2 | 1625 | 143 | ||
GenBank mRNA | BT003242.1 | - | 2 | 1537 | 143 | |
BT133200.1 | - | 2 | 1597 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |